Learn More
Electron Microscopy VACUTAINER STORAGE BX 200 TUBE DFS Item
Supplier:  Electron Microscopy Sciences 64287
Sample Storage Boxes to accommodate our VACUTAINER Tubes. Made from durable cardboard, medical laboratories, forensic laboratories, and hospitals will find these boxes extremely convenient for storing filled vials. These boxes are suitable for incubators, refrigerators, deepfreeze, and controlled atmosphere applications. Model 200-19-117 holds 200 tubes up to 19mm O.D. x up to 117mmL
Catalog No. 5030473
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions