Learn More
BD Vacutainer™ Glass Blood Collection Tubes with Acid Citrate Dextrose (ACD)
Evacuated glass blood collection tubes for blood banking studies, HLA phenotyping and DNA and paternity testing
$147.90 - $1,123.67
Specifications
For Use With (Application) Blood banking
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Additive Tube Size Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Additive Tube Size Price Quantity & Availability  
0268429
View Documents Promotion Details
BD
364816
6 mL ACD Solution B, 1.0mL 13 x 100 mm
Case of 1000 Each for $1,123.67
Only null left
0268426
View Documents Promotion Details
BD
364606
8.5 mL ACD Solution A, 1.5 mL 16 x 100 mm
Pack of 100 Each for $147.90
Case of 10 Pack for $1,479.00
Only null left
Description
Description
  • Vacutainer™ tubes containing Acid Citrate Dextrose (ACD) are used for blood banking studies, HLA phenotyping and DNA and paternity testing
  • ACD Solution A contains trisodium citrate (22.0g/L), citric acid (8.0g/L) and dextrose (24.5g/L)
  • ACD Solution B contains trisodium citrate (13.2g/L), citric acid (4.8g/L) and dextrose (14.7g/L)
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Specifications
Specifications
Blood banking