Learn More
Greiner Bio-One VACUETTE™ Urine CCM Tubes
Description
- Count control and minnitol powder: Additive stabilizes the bacterial count and parameters for up to 48 hours at room temperature.
- Stabilizer contains boric acid, sodium tetraborate, socium formate and mannitol
- Urine specimen does not require refrigeration.
- Suitable to extended processing and transport times, and for microbiological diagnosis
- Leak-proof
- PET plastic is virutally unbreakable
- Feature a vacuum for an optimized sample-to-stabilizer ratio
Specifications
Specifications
| Product Type | Urine Tube |
| Closure Color | Yellow with Black Ring |
| Sterility | Sterile |
| Bottom Shape | Conical |
| Description | Non-ridged |
| No. per Case | 1200/Cs. |
| No. per Pack | 50 Pk. |
| Volume (Metric) | 9 mL |