missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One VACUETTE™ EVOPROTECT SAFETY Blood Collection Set and Luer Adapter

Catalog No. 22405164
Change view
Click to view available options
Needle Gauge:
21 G
23 G
25 G
Length (English) Tube:
7.5 in.
12 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Needle Gauge Length (English) Tube
22405164 23 G 12 in.
22405172 21 G 7.5 in.
22405165 21 G 12 in.
22405173 23 G 7.5 in.
22405174 25 G 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 22405164 Supplier Greiner Bio-One Supplier No. GBO450130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Blood collection set with luer adapter is not made with natural rubber latex or DEHP

  • Blood collection set
  • Luer adapter
  • Single packed
  • Sterile
  • 0.75 in. tubing

Specifications

Product Type Safety Blood Collection Set with Luer Adapter
Color Blue
Needle Gauge 23 G
Sterility Sterile
For Use With (Application) Blood collection
Includes Luer adapter
Length (English) Tube 12 in.
Quantity 60 Pk.