missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHEMetrics Vacu-vials Test Kit, Phenols 1
Description
Snap-and-read Vacu-vials self-filling ampules are the fastest, easiest reagent systems available for precision colorimetric determination of water/wastewater
- Each vacuum-sealed ampule contains a unit dose of premeasured reagent
- Simply immerse the ampule in the water sample, snap its tip, and an analytically correct amount of sample is pulled inside and mixed
- Sample is now ready to be analyzed by your spectrophotometer (instrument must accept a 13 mm cell directly or with an adapter)
- Direct readouts in concentration may be obtained by using CHEMetrics V-2000 Photometer or by preprogramming your spectrophotometer using standard solutions or the data pairs provided in the Vacu-vials instructions
- For immediate results in your spectrophotometer without programming, use supplied equation or calibration tables to convert from absorbance or transmittance to concentration units
Specifications
Specifications
| Analytes | Phenol 1 |
| For Use With (Equipment) | V-2000 photometer or spectrophotometer |
| Includes | Cardboard box, 30 ampoules, 25 mL sample cup, ampoule blank, instructions |
| No. of Tests | 30 |
| Range | 0 to 8 ppm |
| Test Method | 4-Aminoantipyrine |
| Test Time | 1 min. |
| Type | Vacu-vials Instrumental Test Kit |