missing translation for 'onlineSavingsMsg'
Learn More

CHEMetrics Vacu-vials Test Kit, Copper (0 to 7 ppm)

Use with any CHEMetrics Direct-Readout Photometer or spectrophotometer that accepts a 13 mm cell

Supplier:  CHEMetrics K3503

Catalog No. 13-299-309


Only null left

Description

Description

Snap-and-read Vacu-vials self-filling ampules are the fastest, easiest reagent systems available for precision colorimetric determination of water/wastewater

  • Each vacuum-sealed ampule contains a unit dose of premeasured reagent
  • Simply immerse the ampule in the water sample, snap its tip, and an analytically correct amount of sample is pulled inside and mixed
  • Sample is now ready to be analyzed by your spectrophotometer (instrument must accept a 13 mm cell directly or with an adapter)
  • Direct readouts in concentration may be obtained by using CHEMetrics V-2000 Photometer or by preprogramming your spectrophotometer using standard solutions or the data pairs provided in the Vacu-vials instructions
  • For immediate results in your spectrophotometer without programming, use supplied equation or calibration tables to convert from absorbance or transmittance to concentration units

Specifications

Specifications

Copper
30 ampoules, 25 mL sample cup, ampoule blank and instructions
V to 2000: 0 to 12.00 ppm; Spec: 0 to 7.00 ppm
Vacu-vials Instrumental Test Kit
V-2000 photometer or spectrophotometer
30
Bathocuproine
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions