missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vaccine/Cryogenic Vial Gripper
GREENER_CHOICE

Catalog No. 17100483
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17100483 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17100483 Supplier Fisherbrand™ Supplier No. 120910F2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The perfect Vaccine/Cryogenic grip for optimal precision

  • These two styles of hand-operated Grippers are designed to pick up cryogenic and crimped 10R-size vaccine/serum vials, respectively
  • Both are durably made of reinforced polypropylene which is cold-tolerant and resistant to everyday wear
  • The designated squeeze target ensures maximum precision and grip with every use
  • Each style is available in a pack of 5 assorted colors for easy identification and organization
  • A combo pack with one of each style is also available

Specifications

Type Vial and Vaccine Grippers
Color Blue
For Use With (Equipment) Cryogenic Vials and Vaccine Vials