Learn More
Chemglass Inc Vacuum Trap, Serrated Hose Connections, Complete, Trap Bottle 54mm OD x 275mm Overall Length, 50/50 Jt.
Catalog No. CG451504
Encompass
Vacuum Trap, Serrated Hose Connections, Complete, Trap Bottle 54mm OD x 275mm Overall Length, 50/50 Jt. Two piece construction for easy removal of condensate. Inlet and outlet are 180 apart for installation into a variety of vacuum systems. Stopper has a standard taper inner joint and body has an outer joint. Body length is the overall length from the top of the joint to the bottom of the round bottom. Serrated hose connections have an O.D. of 13mm at the smallest serration and 16mm at the largest serration.
Catalog No. CG451504 Supplier Chemglass Inc Supplier No. CG451504
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Vacuum Trap, Serrated Hose Connections, Complete, Trap Bottle 54mm OD x 275mm Overall Length, 50/50 Jt. Two piece construction for easy removal of condensate. Inlet and outlet are 180 apart for installation into a variety of vacuum systems. Stopper has a standard taper inner joint and body has an outer joint. Body length is the overall length from the top of the joint to the bottom of the round bottom. Serrated hose connections have an O.D. of 13mm at the smallest serration and 16mm at the largest serration.