Learn More
Lonza Walkersville Inc SE Cell Line 96-well Nucleofector Kit
Catalog No. NC0103617
Encompass
The SE Cell Line 96-well Nucleofector Kit is one of our three kits suited for medium-throughput transfection of cell lines when working with the 4D-Nucleofector 96-well Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCompatible with liquid handling systems for automated high-throughput electroporationConditions are transferable between 4D Nucleofector System, 4D-Nucleofector 96-well Unit and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10 to 1 x 10 cellsHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:2.25 mL SE Cell Line Nucleofector Solution0.5 mL Supplement 150 g pmaxGFP Vector (1 g/l in 10 mM Tris pH 8.0)1 x 96-well Nucleocuvette Plate (20 L)
Catalog No. NC0103617 Supplier Lonza Walkersville Inc Supplier No. V4SC1096
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The SE Cell Line 96-well Nucleofector Kit is one of our three kits suited for medium-throughput transfection of cell lines when working with the 4D-Nucleofector 96-well Unit.Benefits:High transfection efficiencies in difficult-to-transfect cell linesCompatible with liquid handling systems for automated high-throughput electroporationConditions are transferable between 4D Nucleofector System, 4D-Nucleofector 96-well Unit and 384-well Nucleofector SystemApplications:Transfection of low cell numbers in the range of 5 x 10 to 1 x 10 cellsHigh-throughput applications, e.g. screens of arrayed cDNA, RNAi or CRISPR librariesContent:2.25 mL SE Cell Line Nucleofector Solution0.5 mL Supplement 150 g pmaxGFP Vector (1 g/l in 10 mM Tris pH 8.0)1 x 96-well Nucleocuvette Plate (20 L)