Learn More
MSA™ V-Gard™ Visor
Description
- Thickness helps prevent pitting and warping from heat or weld spatter
- Suitable for applications where welding, cutting and brazing hazards exist
- V-Gard polycarbonate shaded visors are thick and molded to protect against damage from impact, elevated temperatures, weld spatter and infrared (IR) radiation
- Ideal for autogenous welding, metal pouring, gas soldering and light cutting/brazing, they are engineered for heavy-duty use
Specifications
Specifications
| Lens Coating | Anti-scratch, Anti-fog, Impact-resistant |
| Lens Tint | IR Shade 12 |
| Product Type | Visor |
| Material | Polycarbonate |
| Disposable | Reusable |
| Lens Material | Polycarbonate |
| Frame Material | Polycarbonate |
| UV Protection | None |