Learn More
MSA™ V-Gard™ Slotted Full-Brim Hats
Catalog No. 19058247
Change view
Click to view available options
Suspension Type:
Staz-On suspension
Color:
White
Catalog No. Suspension Type Color
19058247 Staz-On suspension White
1 options
Catalog No. 19058247 Supplier MSA™ Supplier No. 454733
Full-brim hardhat available in multiple, fade-resistant colors
  • Polyethylene shell provides superior impact protection
  • Self-adjusting crown straps ensure a comfortable fit
  • Lightweight and well-balanced for all-day wearing comfort
  • Slotted design works with MSA accessories
  • Standard size (6½ to 8)
Suspension System Options:
  • 1-Touch™ Suspension
    • One-handed squeeze opening
    • One-hand slide adjustment
    • Snap closure
    • Adjustable dual nylon straps
  • Staz-On Suspension  
    • 4-point suspension system helps keep helmet in place
    • Features high-quality sweatband and nylon height adjusters
    • Easily clips into V Gard hat or cap shell
    • Adjustable liner
  • Fas-Trac III Suspension
    • Flush rear lug attachments eliminate pressure points
    • Three levels of nape strap adjustment for customized fit
    • Graspable ratchet with ultra-smooth rotation and steadfast hold
Specifications
Color White
Size Standard
Suspension Type Staz-On suspension