Learn More
MSA™ V-Gard™ Protective Slotted Cap with Staz-On™ Suspension
Catalog No. 19080145
Change view
Click to view available options
Color:
Brown
Gray
White
Size:
Standard
Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19080145 Brown Standard
19058250 Gray Standard
1899950 White Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19080145 Supplier MSA™ Supplier No. 464658
Lightweight for all-day wearing comfort
  • Four-point webbed Staz-On suspension with perforated vinyl sweatband
  • Sizes: Small (6-7 1/8), Standard (6 1/2-8), and Large (7-8 1/2)
  • Available in more than 20 colors, with custom imprinting and striping available
Order Info
Contact your Fisher Safety Customer Safety Representative for ordering information for sizes and colors not shown.
†With lamp bracket and cord holder (Class C)
Specifications
Certifications/Compliance Meets or exceeds ANSI Z89.1-2009; Type I; Class E; SEI certified.
Color Brown
Size Standard
Suspension Type Staz-On Pin-Lock