Learn More
BUCHI V-300 Vacuum Pump
Catalog No. 05001090
Change view
Click to view available options
Includes:
Secondary Cold Trap Condenser
Secondary Condenser
Woulff Bottle, Secondary Cold Trap Condenser
Woulff Bottle, Secondary Cold Trap Condenser, I-300 Pro interface
Woulff bottle, Secondary condenser, I-300 Pro interface
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
05001090 Woulff Bottle, Secondary Cold Trap Condenser, I-300 Pro interface
05001080 Secondary Condenser
05001081 Secondary Cold Trap Condenser
05001084 Woulff Bottle, Secondary Cold Trap Condenser
05001089 Woulff bottle, Secondary condenser, I-300 Pro interface
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 05001090 Supplier BUCHI Supplier No. 11V300320
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A quiet and chemically resistant PTFE diaphragm pump. It is optimally suited to work with the I-300 / I-300 Pro Controllers for Rotavapors and parallel evaporators SyncorePlus and Multivapor.
  • Quiet operation thanks to speed regulation
  • Unattended operation in combination with Interface I-300 / I-300 Pro when combined with optional level sensor for secondary condenser that alerts potential overflow of the receiving flask
  • High chemical resistance due to the use of high quality materials
  • Increased lifetime thanks to speed-controlled operation in conjunction with the optional Interface I-300 / I-300 Pro
  • Easy visual detection of condensate and particles due to transparent front panel
  • Seamless plug and play integration into:
    • A speed-controlled vacuum system with Interface I-300 / I-300 Pro
    • A fully automated BUCHI Rotavapor R-300 system including all process parameters
Specifications
Max. Vacuum 5 mbar
Control I-300 Pro Interface
Includes Woulff Bottle, Secondary Cold Trap Condenser, I-300 Pro interface
Flow Rate 1.8 m³/h
Pump Type Diaphragm Pump
Enclosure Rating IP20
Applications Evacuating distillation apparatus, vacuum filtration, vacuum-drying cabinets, drying kilns
Maximum Pressure 2 bar
For Use With (Equipment) I-300/I-300 Pro and R-300
Voltage 100/240 V