Learn More
BUCHI V-300 Vacuum Pump
A quiet and chemically resistant PTFE diaphragm pump. It is optimally suited to work with the I-300 / I-300 Pro Controllers for Rotavapors and parallel evaporators SyncorePlus and Multivapor.
Supplier:  BUCHI 11V300020
 View more versions of this product
Catalog No. 05001081
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Quiet operation thanks to speed regulation
  • Unattended operation in combination with Interface I-300 / I-300 Pro when combined with optional level sensor for secondary condenser that alerts potential overflow of the receiving flask
  • High chemical resistance due to the use of high quality materials
  • Increased lifetime thanks to speed-controlled operation in conjunction with the optional Interface I-300 / I-300 Pro
  • Easy visual detection of condensate and particles due to transparent front panel
  • Seamless plug and play integration into:
    • A speed-controlled vacuum system with Interface I-300 / I-300 Pro
    • A fully automated BUCHI Rotavapor R-300 system including all process parameters
Specifications
Chemical Identifiers
5 mbar
Diaphragm Pump
Evacuating distillation apparatus, vacuum filtration, vacuum-drying cabinets, drying kilns
I-300/I-300 Pro and R-300
1.8 m³/h
IP20
2 bar
100/240 V
Specifications
5 mbar
Secondary Cold Trap Condenser
Diaphragm Pump
Evacuating distillation apparatus, vacuum filtration, vacuum-drying cabinets, drying kilns
I-300/I-300 Pro and R-300
Manual Vacuum Control
1.8 m³/h
IP20
2 bar
100/240 V
Product Suggestions
Product Suggestions