missing translation for 'onlineSavingsMsg'
Learn More

BUCHI V-300 Vacuum Pump

A quiet and chemically resistant PTFE diaphragm pump. It is optimally suited to work with the I-300 / I-300 Pro Controllers for Rotavapors and parallel evaporators SyncorePlus and Multivapor.

Supplier:  BUCHI 11V300100

 View more versions of this product

Catalog No. 05001082


Only null left

Description

Description

  • Quiet operation thanks to speed regulation
  • Unattended operation in combination with Interface I-300 / I-300 Pro when combined with optional level sensor for secondary condenser that alerts potential overflow of the receiving flask
  • High chemical resistance due to the use of high quality materials
  • Increased lifetime thanks to speed-controlled operation in conjunction with the optional Interface I-300 / I-300 Pro
  • Easy visual detection of condensate and particles due to transparent front panel
  • Seamless plug and play integration into:
    • A speed-controlled vacuum system with Interface I-300 / I-300 Pro
    • A fully automated BUCHI Rotavapor R-300 system including all process parameters

Certifications

CE/CSA/CUS

Warranty

1 Year

Specifications

Chemical Identifiers

1.8 m³/hr.
Diaphragm Pump
100/240 V

Specifications

5 mbar (±2 mbar)
Vacuum Pump V-300, Woulff Bottle
1.8 m³/hr.
Rotary evaporation
Manual
PTFE
Diaphragm Pump
100/240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions