Learn More
Invitrogen™ TrueGuide™ gRNA
Ready-to-use, synthetic gRNAs maximize and simplify the performance of CRISPR-Cas9 genome editing experiments.
$145.00
Specifications
| Content And Storage | 1 dried RNA oligo, store at room temperature |
|---|---|
| RNAi Type | gRNA |
| Product Line | TrueGuide |
| Product Type | gRNA |
| Quantity | 2 nmol |
Description
Ready-to-use, synthetic gRNAs maximize the performance of CRISPR-Cas9 genome editing experiments.Specifications
| 1 dried RNA oligo, store at room temperature | |
| TrueGuide | |
| 2 nmol |
| gRNA | |
| gRNA | |
| Dried RNA |
For Research Use Only. Not for use in diagnostic procedures.