Learn More
Carson™ UVSight PRO UV Flashlight with Carabiner Clip Attachment
UVSight PRO UV flashlight contains nine ultra violet LED bulbs, emits 380 nm wavelength for florescent glow
Supplier: Carson™ SL44
Description
- Compact, lightweight flashlight
- Includes two brightness settings
- Glows in dark after being in sunlight
- Powered by 3 AAA batteries (not included)
- For detecting pet stains, auto leaks, counterfeit currency/passports, fake IDs, minerals, gemstones, forensics, scorpions, insects, reptiles, and resin curing
- Backed by Carson™'s one year limited warranty
Includes:
- Carabiner clip attachment for belt loops, backpacks, and key rings
Greener Choice Claims
- Environmental benefits include:
- Free-of a substance that causes environmental harm
- Non-Toxic
Specifications
0.15 lb. | |
68 g | |
Blue | |
UV Flashlight |