missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP UVX Radiometer: UVX-31 Sensor
Description
- The UVX's range switch provides selection from three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 microW/cm2 and 0 to 200 microW/cm2
- A port for connecting a chart recorder (not included) allows for continuous monitoring
- Conversion rate: 2.8 readings/second
- Display: 3.5 digit LCD
- Accuracy: ±2%
- Sensors Accuracy: ±5% (includes NIST standard)
- Dimensions: 6.2L x 3.6W x 2 in.H (157 x 91 x 51mm)
UVX Radiometer: Powered by a 9-volt battery for up to 120 hours
10:1 attenuator: Extends the reading range up to 200mW/cm2
Sensors sold separately.
Quality control
Specifications
Specifications
| Type | UVX-31 Sensor |
| For Use With (Equipment) | UVX Radiometer UVP97001502 |
| Wavelength | 302nm |