missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP UVX Radiometer: UVX-25 Sensor
Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths
Supplier: UVP 97001601
Description
- The UVX's range switch provides selection from three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 microW/cm2 and 0 to 200 microW/cm2
- A port for connecting a chart recorder (not included) allows for continuous monitoring
- Conversion rate: 2.8 readings/second
- Display: 3.5 digit LCD
- Accuracy: ±2%
- Sensors Accuracy: ±5% (includes NIST standard)
- Dimensions: 6.2L x 3.6W x 2 in.H (157 x 91 x 51mm)
UVX Radiometer: Powered by a 9-volt battery for up to 120 hours
10:1 attenuator: Extends the reading range up to 200mW/cm2
Sensors sold separately.
Quality control
Specifications
| UVX-25 Sensor | |
| UVX Radiometer UVP97001502 | |
| 254nm |