missing translation for 'onlineSavingsMsg'
Learn More

UVP UVX Radiometer: Attenuator

Digital UVX Radiometer connects to one of the three interchangeable sensors (sold separately) to measure 254nm, 365nm or 302nm UV wavelengths.

Supplier:  UVP 98003501

Catalog No. UVP98003501


Only null left

Description

Description

  • The UVX's range switch provides selection from three intensity ranges: 0 to 20 mW/cm2, 0 to 2000 microW/cm2 and 0 to 200 microW/cm2
  • A port for connecting a chart recorder (not included) allows for continuous monitoring
  • Conversion rate: 2.8 readings/second
  • Display: 3.5 digit LCD
  • Accuracy: ±2%
  • Sensors Accuracy: ±5% (includes NIST standard)
  • Dimensions: 6.2L x 3.6W x 2 in.H (157 x 91 x 51mm)

UVX Radiometer: Powered by a 9-volt battery for up to 120 hours
10:1 attenuator: Extends the reading range up to 200mW/cm2

Sensors sold separately.

Quality control

Specifications

Specifications

Attenuator, 10:1
Extends reading range up to 200mW/cm2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions