missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVP SI-950 Sterilizing Standard Incubator, 26.9 L, Stainless Steel
Temperature Range: 68°C Max.
Supplier: UVP 95035801
Description
Ideal for incubation of biological assays, fungal, bacterial cultures, eggs and other biological samples.
- Sterilize the chamber between experiments with 254nm shortwave UV to prevent cross contamination
- View samples without opening the door
- Capacity: 0.95 cu. ft. (26.9L)
- Maximum temperature: 68°C, with ±1°C accuracy and ±0.5°C uniformity at 37°C
- Microprocessor-controlled temperature ensures uniformity throughout the chamber
- With two ventilated shelves, adjustable at three positions
- Small footprint—less than 3 sq. ft.—takes up minimal benchtop space
Order Info
Dimensions: 20D x 10.5W x 16.2 in. H (50.8 x 26.7 x 41.1cm)
Specifications
| 0.95 cu. ft. | |
| 26.9 L | |
| 2 | |
| Standard Incubator | |
| 18 in. | |
| 14.75 in. | |
| 60 Hz |
| Microprocessor | |
| Stainless Steel | |
| Ambient to 68°C | |
| ±0.5°C | |
| 45.7 cm | |
| 37.5 cm |