Learn More
UVP Shields for Pen-Ray™ Lamps
Catalog No. UVP98000801
Change view
Click to view available options
Product Type:
Shield A
Catalog No. Product Type
UVP98000801 Shield A
1 options
Catalog No. UVP98000801 Supplier UVP Supplier No. 98000801
Only null left
For directing or modifying the emission output of the 11SC-1 Lamp (UVP90 001 201).
For directing or modifying the emission output of the 11SC-1 Lamp (UVP90 001 201)
  • Shield A has a 0.04 in. I.D. hole for a point-like source
  • Shield B has 0.31 x 0.63 in. window
  • Shield C has 0.19 x 1.5 in. window
Specifications
For Use With (Equipment) Model 11SC-1 Pen-Ray™ UV lamp
Packaging Quantity 1 Ea.
Product Type Shield A