Learn More
UVP J-Series Photovoltaic UV Intensity Meters: Shield Light Attenuator
For use with Photovoltaic J-Series UV meters
Supplier: UVP 23000801
Description
All meters and sensors are individually serialized for traceability of calibration. Calibration is recommended every six months and is certified to meet UVP's published standards and is traceable to NIST. Calibrations are performed at UVP's factory.
Quality control
Greener Choice Claims
- Environmental benefits include:
- Recyclable
Specifications
Attenuator | |
Photovoltaic J-221 and J-225 UV intensity meters |