Learn More
UVP Blak-Ray™ B-100AP High-Intensity UV Inspection Lamps: Exposure Box
Catalog No. UVP76005501
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
UVP76005501 1 Ea.
1 options
Catalog No. UVP76005501 Supplier UVP Supplier No. 76005501
Only null left
For use with Blak-Ray™ Model B-100AP/R lamp
  • Made of heat resistant plastic
  • Cool-Touch™ housing allows users to handle lamp head for a long time
  • Features 100 watt longwave (365nm) bulb and ballast base
  • At 18 in. (457mm), it produces highly intense center of UV irradiance approximately 5 in. (127mm) in diameter
  • Lamp head rotates 360° when attached to base
  • Grip handle fits into the transformer base for easy lamp storage and hands-free lamp operation
Specifications
For Use With (Application) Fluorescing small samples
For Use With (Equipment) Blak-Ray™ Model B-100AP/R lamp
Packaging Quantity 1 Ea.
Product Type Exposure Box