Learn More
Honeywell™ Uvex™ Ultraspec™ 2001 OTG Safety Glasses
Catalog No. 19034242
Change view
Click to view available options
Lens Coating:
Anti-fog
Lens Tint:
Clear
Catalog No. Lens Coating Lens Tint
19034242 Anti-fog Clear
1 options
Catalog No. 19034242 Supplier Honeywell™ Supplier No. UV S0112C
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Comfortable fit over prescription eyewear
These lightweight, unilens spectacles with built-in browguard fit easily over prescription eyewear. Adjustable length Duoflex™ temples provide less pressure behind the ear for improved comfort. In addition, both clear and gray tinted lenses are available with Uvextreme™ coating that resists scratching, fogging, and static. Gray lens is appropriate for outdoor wear; both clear and gray lenses absorb 99.9% of UV radiation up to 400nm.
Order Info
Spectacles weigh 51g.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2015 standards.
Specifications
Lens Coating Anti-fog
Lens Type Uvextreme Anti-fog
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2010
Lens Tint Clear
Product Type Eyewear
Temple Style Duoflex
Weight (Metric) 51 g
UV Protection UV400