Learn More
Honeywell™ Uvex™ Stealth™ OTG Goggles
Catalog No. 19085540
Change view
Click to view available options
Lens Tint:
Clear
Frame Material:
Polycarbonate
Lens Coating:
Anti-fog
Anti-fog, Scratch-resistant
For Use With (Application):
Ideal for most indoor work applications
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Material Lens Coating For Use With (Application)
19085540 Clear Polycarbonate Anti-fog, Scratch-resistant -
296367471 Clear - Anti-fog Ideal for most indoor work applications
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19085540 Supplier Honeywell™ Supplier No. S3970HS
Low-profile, wraparound style provides complete coverage designed for all-day wear
  • Ideal for impact, dust/airborne particles, chemical splash and optical radiation (UV and welding/heat radiation)
  • Superior optics and panoramic field of vision enhanced with toric lens
  • Pivoting clip and quick-adjusting wide headband
  • Elastomer body conforms and contours for a secure fit—even over most prescription eyewear
  • Replacement lenses easily snap on
Order Info
Replacement parts and accessories available (see Cat. No.18-813-429).
Compliance
Meets ANSI Z87.1+ 2015 and CSA Z94.3 standards.
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Frame Material Polycarbonate