Learn More
Honeywell™ Uvex™ Genesis™ Reading Magnifier Safety Glasses
Catalog No. 18813390
Change view
Click to view available options
Lens Type:
Ultra-dura Scratch-resistant
Lens Tint:
Clear
Gray
Magnification Power:
+1.00
+1.50
+2.00
+2.50
+3.00
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Type Lens Tint Magnification Power
18813390 Ultra-dura Scratch-resistant Clear +1.00
18813402 Ultra-dura Scratch-resistant Gray +2.00
18813391 Ultra-dura Scratch-resistant Clear +1.50
18813392 Ultra-dura Scratch-resistant Clear +2.00
18813393 Ultra-dura Scratch-resistant Clear +2.50
18813394 Ultra-dura Scratch-resistant Clear +3.00
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18813390 Supplier Honeywell™ Supplier No. S3760
High-impact protection
  • Soft, flexible nasal fingers
  • Padded temple tips and cushioned browbar
  • Adjustable temple length and ratcheting lens inclination provides for secure fit
  • Easy and economical lens replacement (see Cat. No. 18-813-424)
  • With Ultra-dura™ antiscratch coating
  • Five diopter strengths available
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards.
Specifications
Lens Coating Scratch-resistant
Lens Type Ultra-dura Scratch-resistant
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3-2007
Lens Tint Clear
Product Type Reading Magnifier Spectacles
For Use With (Application) Ideal for most indoor work applications
Frame Color Black
Magnification Power +1.00
UV Protection UV400