Learn More
Honeywell™ Uvex™ Carbonvision™ Safety Goggles
Catalog No. 19111202
Change view
Click to view available options
Frame Material:
Neoprene
Lens Tint:
Clear
Catalog No. Frame Material Lens Tint
19111202 Neoprene Clear
1 options
Catalog No. 19111202 Supplier Honeywell™ Supplier No. S1650D
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal for protection from impact, dust/airborne particles as well as chemical splash hazards
Weighing in at just over an ounce and a half, Uvex Carbonvision goggles are one of the lightest wide vision goggles on the market. Now wearers can work for longer periods without the distractions and problems caused by discomfort, thereby increasing productivity and promoting a safer workplace.
Key Features and Benefits
  • Flexible soft frame adapts perfectly to the face to ensure a perfect fit and seal around the eyes for high-performance protection and all-day comfort
  • Ideal for protection from impact, dust/airborne particles as well as chemical splash hazards
  • Meets the ANSI Z87.1-2010 standard
  • Certified to the requirements of the CSA Z94.3 standard
For more information, visit www.fishersci.com and search for keywords UVEX Goggles.
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Type Dura-streme HC/AF
Certifications/Compliance ANSI Z87.1-2010 and CSA Z94.3 standards
Lens Tint Clear
Product Type Safety Goggles
Frame Color Black, Gray
Frame Material Neoprene