Learn More
Honeywell Safety Products™ Uvex™ Astro OTG 3001™ Safety Glasses
Catalog No. 19084781
Change view
Click to view available options
Frame Color:
Black
Red, White, Blue
Lens Type:
Ultra-dura Hardcoat
Uvextreme Anti-fog
Lens Tint:
Clear
IR Shade 3
IR Shade 5
Lens Coating:
Anti-fog
Scratch-resistant
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Type Lens Tint Lens Coating
19084781 Red, White, Blue Uvextreme Anti-fog Clear Anti-fog
19039134 Black Ultra-dura Hardcoat IR Shade 3 Scratch-resistant
18029 Black Ultra-dura Hardcoat Clear Scratch-resistant
18560B Black Ultra-dura Hardcoat IR Shade 5 Scratch-resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19084781 Supplier Honeywell Safety Products™ Supplier No. S2530C
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Over-the-glasses style
  • Patented adjustable lens inclination and adjustable temple length offer one-size-fits-all flexibility; Duoflex™ temples have soft comfort cushions for the sensitive mastoid area behind the ear
  • Lightweight, impact-resistant nylon frames feature polycarbonate sideshields and browguard for protection against UV, dust, and flying particles
  • Clear polycarbonate lenses with Ultradura™ hardcoat (UD) that resists scratches and absorbs 99.9% of UV radiation up to 385nm
uvextreme™ Anti-Fog Coating (XTR)
  • Permanently bonded to lens
  • Absorbs 99.9% of UV radiation
  • Anti-scratch and -static
  • Will not wear after repeated cleanings
ultra-dura™ Hard Coat (UD)
  • Permanently bonded to lens
  • Scratch- and chemical-resistant
Order Info
Customization with a company name, logo, or department is available. For details, call your Fisher Scientific Customer Service Representative at 1-800-772-6733.
Compliance
Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards. Meet stringent military V0 Ballistic Test for Impact requirements.
Specifications
Lens Coating Anti-fog
Lens Type Uvextreme Anti-fog
Certifications/Compliance ANSI Z87.1-2015, CSA Z94.3, Military V0 Ballistic Test
Description Clear XTR
Lens Tint Clear
Product Type Over-the-Glasses Safety Spectacles
Frame Color Red, White, Blue
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400