missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One UV-Star™ 96-well UV Spectroscopy Microplate
GREENER_CHOICE

Catalog No. 07000724
Change view
Click to view available options
Material:
Cycloolefin Copolymer
Polystyrene
Well Design:
Chimney
Half area
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Well Design
07000724 Polystyrene Half area
07000407 Cycloolefin Copolymer Chimney
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07000724 Supplier Greiner Bio-One Supplier No. 675801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in UV Spectroscopy. Greiner Bio-One UV-Star™ 96-Well UV Spectroscopy Microplate is used for measurements of protein concentration at 280nm.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Can be traced back to production through defined LOT number system
  • Optical window down to 230nm ideal for nucleic acid

Service Plan

No

Warranty

No

Specifications

Barcode Alphanumeric
Breakapart No
Format Half-Area
Array 8 x 12
Packaging Inner bags of 10
Binding Type Non Binding
Color Clear
Product Type UV Microplate
Material Polystyrene
Volume (Metric) Well 15 to 175 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 199 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) UV Spectroscopy
Quantity 40/Cs.
Well Design Half area
Dimensions (L x W x H) 127.8 x 85.5 x 11.5 mm
Show More Show Less