missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One UV-Star™ 96-well UV Spectroscopy Microplate
GREENER_CHOICE

Catalog No. 07000407
Change view
Click to view available options
Material:
Cycloolefin Copolymer
Polystyrene
Well Design:
Chimney
Half area
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Well Design
07000407 Cycloolefin Copolymer Chimney
07000724 Polystyrene Half area
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07000407 Supplier Greiner Bio-One Supplier No. 655801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in UV Spectroscopy. Greiner Bio-One UV-Star™ 96-Well UV Spectroscopy Microplate is used for measurements of protein concentration at 280nm.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Can be traced back to production through defined LOT number system
  • Optical window down to 230nm ideal for nucleic acid

Specifications

Breakapart No
Format Standard
Lid Without Lid
Array 8 x 12
Packaging Inner bags of 10
Binding Type Non Binding
Color Clear
Product Type 96-well Microplate
Material Cycloolefin Copolymer
No. of Wells 96
Sterility Non-sterile
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) UV Spectroscopy
Quantity 40/Cs.
Well Design Chimney
Dimensions (L x W x H) 127.8 x 85.5 x 11.5 mm
Show More Show Less