missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One UV-Star™ 384-well Microplate Flat Bottom Microplate
GREENER_CHOICE

Catalog No. 07000406
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07000406 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07000406 Supplier Greiner Bio-One Supplier No. GB781801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for measuring protein concentration at 280nm.

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Used for nucleic acid determinations at 260nm/280nm
  • DMSO-resistant

Specifications

Barcode No Barcode
Format Standard
Lid Without Lid
Array 16 x 24
Autoclavable Not Autoclavable
Color Clear
Product Type Microplate
Material Cycloolefin Copolymer
No. of Wells 384
Volume (Metric) Working 15 to 110 μL
Sterility Non-sterile
Total Volume (Metric) Well 131 μL
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 10
Quantity 40/Cs.
RNase Free Yes
Temperature Range (Metric) -80° to +40°C
Show More Show Less