Learn More
Perkin Elmer Canada UV Light Shield for ELAN
Catalog No. 509048112
Encompass
The UV light safety shield protects users from rays given off by the plasma in the instrument and secures the spray chamber to the base of the injector. It is included in the Quartz Cyclonic Sample Introduction System for ELAN (N8120519). For the NexION 350 use Part Number N8142047. For the NexION 1000/2000 use Part Number N8152371.
Catalog No. 509048112 Supplier Perkin Elmer Canada Supplier No. W1020996
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The UV light safety shield protects users from rays given off by the plasma in the instrument and secures the spray chamber to the base of the injector. It is included in the Quartz Cyclonic Sample Introduction System for ELAN (N8120519). For the NexION 350 use Part Number N8142047. For the NexION 1000/2000 use Part Number N8152371.