missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Extended Length, Bulk
GREENER_CHOICE

Catalog No. 12111027
Change view
Click to view available options
Volume (Metric):
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12111027 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12111027 Supplier BIOTIX™ Supplier No. 63300077
Only null left

Packaged in re-sealable bags making storage and racking simple. Biotix™ uTIP™ Low Retention Non-Filter Universal Pipette Tips, Bulk are RNase, DNase, Pyrogens, and PCR Inhibitors free.

  • Fits leading pipette brands and improves ergonomics by lowering insertion and ejection forces
  • Achieved through patented technologies including FlexFit and Blade
  • Contain X-Resin giving uniform low retention properties
  • Results in minimal sample retention in tip improving accuracy and precision of results
  • Re-sealable bag
  • Tips must be racked by hand
  • Sterile products undergo E-Beam radiation

Certifications

ISO 9001 , FDA Class I

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Format Bulk
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line uTip
Material Polypropylene
Show More Show Less