missing translation for 'onlineSavingsMsg'
Learn More

BIOTIX™ uTIP™ CleanPak™ Reload, Sterile Low Retention Universal Pipette Tips,Extended Length
GREENER_CHOICE

Catalog No. 12111022
Change view
Click to view available options
Volume (Metric):
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
12111022 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12111022 Supplier BIOTIX™ Supplier No. 63300070
Only null left

Designed with hermetically sealed bag to avoid contamination. Biotix™ uTIP™ CleanPak™ Reload, Low-Retention, Non-Filter Universal Pipette Tips reduce time to rack products, cost compared to pre-racked products, and environmental footprint.

  • Fits all standard pipettes
  • Improves ergonomics by lowering insertion and ejection forces with FlexFit and Blade technologies
  • CleanPak reload features advanced technology of Biotix tips
  • Allows re-use of previous racks
  • 960 tips on lab bench using the same surface area as 1 rack
  • Sterile and non-Sterile re-sealable bag available
  • Clear bag and color coded snap cards for quick identification
  • Fully recyclable and contains no contaminating cardboard or paper
  • Inserts are easy to transfer to Biotix empty racks
  • Sterile products undergo E-Beam radiation
  • All racks include label with unique bar-coded serial number, part number, and description for traceability to individual rack

Certifications

ISO 9001 , FDA Class I

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Refill
Tip Style Fine Point
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line uTIP CleanPak
Material Polypropylene
Show More Show Less