missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Utility Trays for Shaker Platforms

Catalog No. 236027
Change view
Click to view available options
Dimensions (L x W):
45.7 x 45.7 cm
353 x 272 mm
425 x 209 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
236027 45.7 x 45.7 cm
01258076 425 x 209 mm
01258077 353 x 272 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 236027 Supplier Thermo Scientific™ Supplier No. 236027
Only null left

Use these utility trays with 0.5 in. rubber mats with Thermo Scientific™ shakers.

Use these Thermo Scientific™ Utility trays for Shaker Platforms with 0.5 in. rubber mats.

Thermo Scientific™ utility trays for Shaker platforms have a 0.5 rubber mat to suit individual needs in the laboratory.

Specifications

Product Type Utility Tray
Dimensions (L x W) 45.7 x 45.7 cm
Clamps 45.7 cm
Description Utility tray for 18 x 18 in. platform
For Use With (Equipment) MaxQ HP 416, 420, 430, 435, 436, 480, 481