Learn More
StatLab Uterus Control Microscope Slides
Catalog No. 22000026
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
22000026 25 Pk.
1 options
Catalog No. 22000026 Supplier StatLab Supplier No. CSU0325P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Uterus Control Microscope Slides for special stains
  • Uterus Control Slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners
  • Each package includes specific staining protocol
  • 25/package
  • Stained slide is not included
  • Tissue: Uterus
Specifications
Product Type Control Slide
Quantity 25 Pk.