missing translation for 'onlineSavingsMsg'
Learn More

Code Corporation™ USB to Micro USB 3 ft. Cable for Code Reader Barcode Scanner

Catalog No. 17579701
Change view
Click to view available options
For Use With (Equipment):
CR2700 INDUCTIVE CHARGING STATION (151789)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17579701 CR2700 INDUCTIVE CHARGING STATION (151789)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17579701 Supplier Code Corporation™ Supplier No. 176506

Use this 3-foot straight USB cable to make a USB to Micro USB connection.

  • Dark Gray, 3 ft
  • Reader termination: Micro USB
  • Host termination: USB Type A

Specifications

For Use With (Equipment) CR2700 INDUCTIVE CHARGING STATION (151789)
Length (English) 3 ft.
Type Cable
Alternate Part Number CRA-C34-C286