missing translation for 'onlineSavingsMsg'
Learn More

Code Corporation™ USB to Micro USB 6 ft. Cable for Code Reader Barcode Scanner

Catalog No. 17579685
Change view
Click to view available options
Connection Type:
9 pin D-Sub
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
17579685 9 pin D-Sub
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17579685 Supplier Code Corporation™ Supplier No. 176508

Use this 6 ft. straight cable for making USB to Micro USB connections.

  • Reader termination: Micro USB
  • Host termination: USB Type A

Specifications

For Use With (Equipment) CR2700 Inductive Charging Station
Type Cable
Connection Type 9 pin D-Sub
Alternate Part Number CRA-C36-C286