missing translation for 'onlineSavingsMsg'
Learn More

Code Corporation™ USB to RJ50 6 ft. Cable for Code Reader Barcode Scanner

Catalog No. 17579703
Change view
Click to view available options
For Use With (Equipment):
CR900FD, CR1100, CR1500, CR5200, and CR950 Scanners
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17579703 CR900FD, CR1100, CR1500, CR5200, and CR950 Scanners
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17579703 Supplier Code Corporation™ Supplier No. 176507

Use this 6-foot straight cable for making USB to RF50 connections.

  • Dark Gray, 6 ft.
  • Reader termination: RJ50
  • Host termination: USB Type A

Specifications

For Use With (Equipment) CR900FD, CR1100, CR1500, CR5200, and CR950 Scanners
Length (English) 6 ft.
Type Cable
Alternate Part Number CRA-C500-C286