missing translation for 'onlineSavingsMsg'
Learn More

Brady™ USB-A to RJ50 Cable for V2200 Wired Barcode Scanner

Catalog No. 19029967
Change view
Click to view available options
Length (English):
6.56 ft.
9.84 ft.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19029967 9.84 ft.
19029966 6.56 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19029967 Supplier Brady™ Supplier No. 178573
Only null left

For use with the V2200 Wired Barcode Scanner.

Replacement cable for use with the V2200 Wired Barcode Scanner.

Available as 2 m straight cable or 3 m coiled cable.

Specifications

Length (English) 9.84 ft.
Length (Metric) 3 m
Type Cable