missing translation for 'onlineSavingsMsg'
Learn More

Ursolic Acid 90.0+%, TCI America™

Catalog No. U0065100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U0065100MG 100 mg
U00651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U0065100MG Supplier TCI America Supplier No. U0065100MG

Chemical Identifiers

CAS 77-52-1
Molecular Formula C30H48O3
Molecular Weight (g/mol) 456.71
MDL Number MFCD00009621
InChI Key WCGUUGGRBIKTOS-GPOJBZKASA-N
Synonym 3beta-Hydroxy-12-ursen-28-oic Acid
PubChem CID 129316832
IUPAC Name (1S,2R,4aS,6aS,6bR,8aR,10S,12aR,12bR,14bS)-10-hydroxy-1,2,6a,6b,9,9,12a-heptamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylic acid
SMILES C[C@@H]1CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@H]1C)C(O)=O

Specifications

Color White
Quantity 100 mg
Formula Weight 456.71
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ursolic Acid