missing translation for 'onlineSavingsMsg'
Learn More

Urocanic acid, 98%

Catalog No. p-3779864
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228960050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC228960050 Supplier Thermo Scientific Chemicals Supplier No. 228960050
Only null left

CAS: 104-98-3 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 104-98-3
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00005203
InChI Key LOIYMIARKYCTBW-UPHRSURJSA-N
Synonym urocanic acid, 4-imidazoleacrylic acid, trans-urocanic acid, urocaninic acid, imidazoleacrylic acid, 5-imidazoleacrylic acid, urocanate, imidazole-4-acrylic acid, 2e-3-1h-imidazol-4-yl acrylic acid, 3-1h-imidazol-4-yl-2-propenoic acid
PubChem CID 1549103
ChEBI CHEBI:30818
IUPAC Name (Z)-3-(1H-imidazol-5-yl)prop-2-enoic acid
SMILES C1=C(NC=N1)C=CC(=O)O

Specifications

Melting Point 217.0°C to 220.0°C
Color White to Beige
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 25, 126
Merck Index 10, 9691
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,,insoluble in ethanol,diethyl ether
Formula Weight 138.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Urocanic acid
Show More Show Less
EINECSNumber 203-258-4

RUO – Research Use Only