missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Urine System Accessories

For hygienic transfer of urine from collection devices

$42.45 - $42.45

Specifications

Material PET
Certifications/Compliance CE
For Use With (Application) For hygienic transfer of Urine from collection devices
Storage Requirements 4°C to 25°C
View More Specs

Products 1
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
22-040-114
View Documents
Greiner Bio-One
450251
Urine Transfer Device
Pack of 50 Each for $42.45
Case of 12 Pack for $509.40
Only null left
 
Description

Description

Specifications

Specifications

PET
CE
For hygienic transfer of Urine from collection devices
4°C to 25°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'