Learn More
Fisherbrand™ Urine Sedimentation System, Complete Urine Kit and Specimen Transport
Catalog No. 14377201
Change view
Click to view available options
Volume (Metric):
15 mL
Catalog No. Volume (Metric)
14377201 15 mL
1 options
Catalog No. 14377201 Supplier Fisherbrand™ Supplier No. 14377201
Designed to generate the same results as the most popular urine sedimentation system using the same test procedure, the Fisherbrand Urine Sedimentation System features a 15mL graduated urine tube that produces 1mL of urine containing sediment for further evaluation.
  • The urine tubes are available in economical or break-resistant polystyrene (caps are available separately)
  • One Fisherbrand™ UriSystem™ DeciSlide™ can be used to analyze up to 10 samples simultaneously
Specifications
Includes 500 Super Tubes, 500 Cups, 500 Caps, 500 Labels, and 5 Specimen Racks
Tube Size 15 mL
Volume (Metric) 15 mL