missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Urine Container with Integrated Transfer Device

Catalog No. 23555800
Change view
Click to view available options
Volume (Metric):
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23555800 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23555800 Supplier Fisherbrand™ Supplier No. 3856LPLFH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Features an integrated transfer mechanism for closed system transfer of urine samples directly from the cup to a vacuum sample tube.

  • Requires no open transferring of samples for reduced spills, improves lab hygiene, and limited exposure to dangerous samples
  • Reduced risk of contamination for improved test results
  • Directly transfers samples to a tube suitable for analysis and transport
  • Works with most vacuum urine tubes
  • Provides consistent sample transfer volumes when used with like vacuum tubes
  • Improves efficiencies and sample throughput when compared to traditional open systems

Specifications

Closure Color Yellow
Closure Type Screw Cap
Disposable Single-use
Graduated Yes
Material Polypropylene
Sterility Sterile
Color Clear
For Use With (Application) Urine Vacuum Tubes
Includes Integrated Transfer Device
Latex Free Yes
No. per Case 300/Cs.
Volume (English) 4 oz.
Volume (Metric) 120 mL
Show More Show Less