Learn More
Greiner Bio-One Urine Collection Tubes
For collection and transport of urine specimens
Supplier:  Greiner Bio-One 456007
 View more versions of this product
Catalog No. 22040126
Only null left
Description
Description
  • Sterile, leak-proof tubes made of clear, unbreakable disposable plastic
  • Pre-defined vacuum for exact draw volumes
  • Use tubes with round bottoms for chemical analyses; conical bottom tubes for microscopic examination of urine sediment
  • Use tubes with boric acid preservative for specimens tested for an otherwise unstable analyte or specimens for microbiological studies (stable up to 24 hours)
Tubes with preservatives should be inverted 6-8 times following collection in order to achieve homogenous mixing
Specifications
Chemical Identifiers
Urine Tube
Stopper
Round
For microscopic examination of Urine sediment
Yellow
PET
CE
4°C to 25°C
Specifications
Urine Tube
Stopper
Round
For microscopic examination of Urine sediment
0.1 oz.
Yellow
PET
CE
4°C to 25°C
6.5 mL
Product Suggestions
Product Suggestions