missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Uridine-5'-triphosphate trisodium salt, 98+%

Catalog No. p-7042702
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
250 mg
Catalog No. Quantity
AAJ63427MC 100 mg
AAJ63427MD 250 mg
2 options

Catalog No. AAJ63427MC

Supplier: Thermo Scientific Chemicals J63427MC

Only null left

CAS: 19817-92-6 | C9H12N2Na3O15P3 | 550.09 g/mol

Uridine-5'-triphosphate trisodium salt, is used as an energy-rich precursor in the enzymatic biosynthesis of RNA, a potent vasodilator, and induces contractile responses in some tissues. Uridine-5?-triphosphate, Trisodium Salt is an activator of P2Y. UTP has been used to study its vasodilatory effects on the skeletal muscle cells of diabetic patients1. UTP has also been used as a purinergic agonist to study ion transport functions of P2Y2 receptors on rat basolateral membranes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Uridine-5′-triphosphate trisodium salt, is used as an energy-rich precursor in the enzymatic biosynthesis of RNA, a potent vasodilator, and induces contractile responses in some tissues. Uridine-5′-triphosphate, Trisodium Salt is an activator of P2Y. UTP has been used to study its vasodilatory effects on the skeletal muscle cells of diabetic patients1. UTP has also been used as a purinergic agonist to study ion transport functions of P2Y2 receptors on rat basolateral membranes

Solubility
Soluble in water (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 19817-92-6
Molecular Formula C9H12N2Na3O15P3
Molecular Weight (g/mol) 550.09
MDL Number MFCD11045990
InChI Key MMJGIWFJVDOPJF-ZKGVITBDNA-K
Synonym uridine-5'-triphosphate trisodium salt dihydrate, uridine 5'-triphosphate trisodium salt dihydrate, trisodium uridine 5'-triphosphate dihydrate, 550.0866 anhydrous, c9h12n2na3o15p3.xh2o, uridine 5'-tetrahydrogen triphosphate , sodium salt, hydrate 1:3:2, trisodium dihydrate utp trianion, sodium 5'-o-hydroxyphosphinato oxy phosphinato oxy phosphinato uridine hydrate 3:1:2, trisodium, 2r,3s,4r,5r-5-2,4-dioxopyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methoxy-oxidophosphoryl oxy-oxidophosphoryl hydrogen phosphate, dihydrate, uridine 5'-triphosphate, trisodium salt, hydrate
PubChem CID 131675549
IUPAC Name trisodium;(2R,3R,4S,5R)-2-(2,4-dioxopyrimidin-1-yl)-5-[[hydroxy-[hydroxy(phosphonooxy)phosphoryl]oxyphosphoryl]oxymethyl]oxolane-3,4-diolate
SMILES [Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N1C=CC(=O)NC1=O

Specifications

Color White
Quantity 100 mg
Beilstein 3585234
Solubility Information Soluble in water (50mg/ml).
Formula Weight 550.09
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Uridine-5'-triphosphate trisodium salt
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 243-347-5
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only