missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine 5'-Monophosphate Disodium Salt Hydrate 98.0+%, TCI America™
Supplier: TCI America U002125G
Specifications
| Uridine 5′-Monophosphate Disodium Salt Hydrate | |
| White | |
| MFCD00149422 | |
| uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt | |
| [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O | |
| 368.15 | |
| 368.15 | |
| Crystalline Powder |
| 3387-36-8 | |
| C9H11N2Na2O9P | |
| 25 g | |
| KURVIXMFFSNONZ-IJRYJGAHNA-L | |
| disodium [(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate | |
| 123134060 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 3387-36-8 | |
| 368.15 | |
| KURVIXMFFSNONZ-IJRYJGAHNA-L | |
| 123134060 | |
| [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O |
| C9H11N2Na2O9P | |
| MFCD00149422 | |
| uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt | |
| disodium [(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate |
Safety and Handling
EINECSNumber : (8)-0278&(9)-0196
RTECSNumber : YU7975000
TSCA : No
Recommended Storage : Refrigerator