missing translation for 'onlineSavingsMsg'
Learn More

Uridine 5'-Monophosphate Disodium Salt Hydrate 98.0+%, TCI America™

Catalog No. U00215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00215G 5 g
U002125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00215G Supplier TCI America Supplier No. U00215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3387-36-8
Molecular Formula C9H11N2Na2O9P
Molecular Weight (g/mol) 368.15
MDL Number MFCD00149422
InChI Key KURVIXMFFSNONZ-IJRYJGAHNA-L
Synonym uridine-5'-monophosphate disodium salt, uridine 5'-monophosphate, disodium salt, 5'-uridylicaciddisodiumsalt
PubChem CID 123134060
IUPAC Name disodium [(2R,3S,4R,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])([O-])=O)N1C=CC(=O)NC1=O

Specifications

Color White
Quantity 5 g
Formula Weight 368.15
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Uridine 5′-Monophosphate Disodium Salt Hydrate
EINECSNumber (8)-0278&(9)-0196
RTECSNumber YU7975000
TSCA No
Recommended Storage Refrigerator